Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135213.1 | NP_001128685.1 | 16 - 17 | 1744 | 67 |
NM_001135214.1 | NP_001128686.1 | 16 - 17 | 1888 | 67 | |
NM_014776.3 | NP_055591.2 | 15 - 16 | 1738 | 67 | |
NM_057169.3 | NP_476510.1 | 17 - 18 | 1978 | 67 | |
NM_057170.3 | NP_476511.1 | 15 - 16 | 1594 | 67 | |
XM_005253997.3 | XP_005254054.1 | 17 - 18 | 1970 | 67 | |
XM_005254000.3 | XP_005254057.1 | 16 - 17 | 1820 | 67 | |
XM_006719707.3 | XP_006719770.1 | 17 - 18 | 1928 | 67 | |
XM_006719708.3 | XP_006719771.1 | 16 - 17 | 1838 | 67 | |
XM_006719709.3 | XP_006719772.1 | 16 - 17 | 1823 | 67 | |
XM_006719712.3 | XP_006719775.1 | 14 - 15 | 1484 | 67 | |
XM_017020258.1 | XP_016875747.1 | 16 - 17 | 1835 | 67 | |
XM_017020259.1 | XP_016875748.1 | 17 - 18 | 1970 | 67 | |
XM_017020260.1 | XP_016875749.1 | 16 - 17 | 1880 | 67 | |
GenBank mRNA | AF124491.1 | - | 17 - 18 | 1813 | 67 |
AK303198.1 | - | 15 - 16 | 1589 | 67 | |
AK308304.1 | - | 16 - 17 | 1802 | 67 | |
BC014223.2 | - | 14 - 15 | 1345 | 67 | |
CR749340.1 | - | 3 - 4 | 469 | 67 | |
D63482.2 | - | 16 - 17 | 1650 | 67 |