Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001200054.1 | NP_001186983.1 | 2 - 3 | 242 | 68 |
NM_001320121.1 | NP_001307050.1 | 2 - 3 | 242 | 68 | |
NM_001320122.1 | NP_001307051.1 | 2 - 3 | 242 | 68 | |
NM_006928.4 | NP_008859.1 | 3 - 4 | 277 | 68 | |
XM_006719569.1 | XP_006719632.1 | 2 - 3 | 854 | 68 | |
XM_011538685.1 | XP_011536987.1 | 3 - 4 | 306 | 68 | |
GenBank mRNA | AK092881.1 | - | 2 - 3 | 855 | 68 |
AK130180.1 | - | 3 - 4 | 301 | 68 | |
BC001414.2 | - | 2 - 3 | 201 | 68 | |
BT007202.1 | - | 2 - 3 | 191 | 68 | |
DA035117.1 | - | 3 - 4 | 274 | 68 | |
DA633173.1 | - | 3 - 4 | 268 | 68 | |
M32295.1 | - | 2 - 3 | 212 | 68 | |
M77348.1 | - | 2 - 3 | 202 | 68 | |
S73003.1 | - | 2 - 3 | 212 | 68 | |
U01874.1 | - | 2 - 3 | 197 | 68 |