Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281729.1 | NP_001268658.1 | 1 - 2 | 82 | 87 |
NM_001319157.1 | NP_001306086.1 | 1 - 2 | 82 | 87 | |
NM_014183.3 | NP_054902.1 | 1 - 2 | 82 | 87 | |
NR_104032.1 | 1 - 2 | 82 | 87 | ||
GenBank mRNA | BG110161.1 | - | 1 - 2 | 67 | 87 |