Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276380.1 | NP_001263309.1 | 12 - 13 | 2213 | 78 |
NM_016649.3 | NP_057733.2 | 12 - 13 | 2224 | 78 | |
XM_017027874.1 | XP_016883363.1 | 12 - 13 | 3676 | 78 | |
GenBank mRNA | AK074299.1 | - | 11 - 12 | 1710 | 78 |