Assay Design



Gene Transcripts

Gene Symbol
RXRA
Entrez Gene ID
6256
Gene Name
retinoid X receptor alpha
Gene Aliases
NR2B1
Chromosome Location
Chr.9: 134326463 - 134440586 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291920.1 NP_001278849.1 2 - 3 1118 110
NM_002957.5 NP_002948.1 2 - 3 446 110
GenBank mRNA AB307705.1 - 2 - 3 277 110
AK131513.1 - 2 - 3 1118 110
BC063827.1 - 2 - 3 439 110
BC096246.2 - 8 - 9 110
BC110998.1 - 2 - 3 408 110
BQ220698.1 - 2 - 3 446 110
HQ692843.1 - 2 - 3 277 110
X52773.1 - 2 - 3 352 110

View Full Product Details