Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291920.1 | NP_001278849.1 | 3 - 4 | 1272 | 98 |
NM_001291921.1 | NP_001278850.1 | 2 - 3 | 1063 | 98 | |
NM_002957.5 | NP_002948.1 | 3 - 4 | 600 | 98 | |
GenBank mRNA | AB307705.1 | - | 3 - 4 | 431 | 98 |
AK131192.1 | - | 2 - 3 | 1063 | 98 | |
AK131513.1 | - | 3 - 4 | 1272 | 98 | |
BC063827.1 | - | 3 - 4 | 593 | 98 | |
BC096246.2 | - | 7 - 8 | 98 | ||
BC110998.1 | - | 3 - 4 | 562 | 98 | |
BQ220698.1 | - | 3 - 4 | 601 | 99 | |
HQ692843.1 | - | 3 - 4 | 431 | 98 | |
X52773.1 | - | 3 - 4 | 506 | 98 |