Assay Design



Gene Transcripts

Gene Symbol
DPYSL4
Entrez Gene ID
10570
Gene Name
dihydropyrimidinase like 4
Gene Aliases
CRMP3, DRP-4, ULIP4
Chromosome Location
Chr.10: 132184983 - 132205776 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011539125.2 XP_011537427.1 3 - 4 281 76
XM_017015483.1 XP_016870972.1 3 - 4 1695 76

View Full Product Details