Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014510.2 | NP_055325.2 | 19 - 20 | 14888 | 90 |
NM_033026.5 | NP_149015.2 | 19 - 20 | 14888 | 90 | |
XM_017012006.1 | XP_016867495.1 | 17 - 18 | 9068 | 90 | |
XM_017012007.1 | XP_016867496.1 | 16 - 17 | 9025 | 90 | |
GenBank mRNA | AB011131.1 | - | 13 - 14 | 3432 | 90 |
BC001304.1 | - | 10 - 11 | 1094 | 90 | |
BC108289.1 | - | 9 - 10 | 1063 | 90 |