Assay Design



Gene Transcripts

Gene Symbol
PCLO
Entrez Gene ID
27445
Gene Name
piccolo presynaptic cytomatrix protein
Gene Aliases
ACZ, PCH3
Chromosome Location
Chr.7: 82754005 - 83164162 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014510.2 NP_055325.2 5 - 6 9386 82
NM_033026.5 NP_149015.2 5 - 6 9386 82

View Full Product Details