Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014510.2 | NP_055325.2 | 8 - 9 | 13725 | 65 |
NM_033026.5 | NP_149015.2 | 8 - 9 | 13725 | 65 | |
XM_017012006.1 | XP_016867495.1 | 6 - 7 | 7905 | 65 | |
XM_017012007.1 | XP_016867496.1 | 6 - 7 | 7889 | 65 | |
GenBank mRNA | AB011131.1 | - | 2 - 3 | 2269 | 65 |