Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014510.2 | NP_055325.2 | 11 - 12 | 14057 | 154 |
NM_033026.5 | NP_149015.2 | 11 - 12 | 14057 | 154 | |
XM_017012006.1 | XP_016867495.1 | 9 - 10 | 8237 | 154 | |
XM_017012007.1 | XP_016867496.1 | 9 - 10 | 8221 | 154 | |
GenBank mRNA | AB011131.1 | - | 5 - 6 | 2601 | 154 |
BC108289.1 | - | 2 - 3 | 259 | 154 |