Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016234.3 | NP_057318.2 | 3 - 4 | 541 | 105 |
NM_203379.1 | NP_976313.1 | 3 - 4 | 402 | 105 | |
NM_203380.1 | NP_976314.1 | 3 - 4 | 568 | 105 | |
XM_017016324.1 | XP_016871813.1 | 3 - 4 | 576 | 105 | |
GenBank mRNA | AB033899.1 | - | 3 - 4 | 415 | 105 |
AK000339.1 | - | 3 - 4 | 551 | 105 | |
AK024932.1 | - | 3 - 4 | 542 | 105 | |
AK222782.1 | - | 3 - 4 | 559 | 105 | |
AK223288.1 | - | 3 - 4 | 530 | 105 | |
AK310436.1 | - | 3 - 4 | 351 | 105 | |
AM262166.1 | - | 2 - 3 | 261 | 105 | |
AY358520.1 | - | 3 - 4 | 502 | 105 | |
BC007985.2 | - | 3 - 4 | 519 | 105 | |
CF595490.1 | - | 3 - 4 | 405 | 105 |