Assay Design
Gene Transcripts
- Gene Symbol
- STEAP1B
- Entrez Gene ID
-
256227
- Gene Name
- STEAP family member 1B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 22419444 - 22500282 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_207342.2 |
NP_997225.1 |
4 - 5 |
907 |
79 |
GenBank mRNA |
BC066301.1 |
- |
4 - 5 |
804 |
79 |
View Full Product Details