Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005819.5 | NP_005810.1 | 2 - 3 | 649 | 118 |
XM_011509058.1 | XP_011507360.1 | 2 - 3 | 649 | 118 | |
GenBank mRNA | AJ002078.1 | - | 2 - 3 | 201 | 118 |
AK312440.1 | - | 2 - 3 | 436 | 118 | |
BC009944.2 | - | 2 - 3 | 372 | 118 | |
BU902578.1 | - | 2 - 3 | 439 | 118 | |
CR541856.1 | - | 2 - 3 | 201 | 118 | |
CR541873.1 | - | 2 - 3 | 201 | 118 |