Assay Design
Gene Transcripts
- Gene Symbol
- ELMOD2
- Entrez Gene ID
-
255520
- Gene Name
- ELMO domain containing 2
- Gene Aliases
-
9830169G11Rik
- Chromosome Location
-
Chr.4: 140524146 - 140553770 on Build GRCh38
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
MGAT4 family member D |
MGAT4D |
GnT1IP |
-43.9kb
|
Anti-sense |
Chr. - 140442257 - 140499829 on Build GRCh38 |
Downstream |
uncoupling protein 1 |
UCP1 |
SLC25A7, UCP |
9.9kb
|
Anti-sense |
Chr. - 140555770 - 140568914 on Build GRCh38 |
View Full Product Details