Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286734.1 | NP_001273663.1 | 44 - 45 | 7269 | 71 |
NM_001813.2 | NP_001804.2 | 46 - 47 | 7632 | 71 | |
XM_011531544.2 | XP_011529846.1 | 45 - 46 | 7550 | 71 | |
XM_011531545.2 | XP_011529847.1 | 45 - 46 | 7472 | 71 | |
XM_011531546.2 | XP_011529848.1 | 45 - 46 | 7427 | 71 | |
XM_011531547.2 | XP_011529849.1 | 45 - 46 | 7343 | 71 | |
XM_011531548.2 | XP_011529850.1 | 45 - 46 | 7340 | 71 | |
XM_011531549.2 | XP_011529851.1 | 44 - 45 | 7187 | 71 | |
XM_017007659.1 | XP_016863148.1 | 44 - 45 | 7181 | 71 | |
GenBank mRNA | AB209996.1 | - | 44 - 45 | 7194 | 71 |
Z15005.1 | - | 47 - 48 | 7518 | 71 |