Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005956.3 | NP_005947.3 | 14 - 15 | 1801 | 109 |
GenBank mRNA | AK302747.1 | - | 14 - 15 | 1811 | 109 |
AK309266.1 | - | 13 - 14 | 1414 | 109 | |
AK312361.1 | - | 14 - 15 | 1506 | 109 | |
BC009806.2 | - | 14 - 15 | 1467 | 109 | |
BC050420.1 | - | 14 - 15 | 1492 | 109 | |
J04031.1 | - | 14 - 15 | 1467 | 109 |