Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005956.3 | NP_005947.3 | 21 - 22 | 2523 | 76 |
GenBank mRNA | AK302747.1 | - | 21 - 22 | 2533 | 76 |
AK312361.1 | - | 21 - 22 | 2228 | 76 | |
BC001014.2 | - | 4 - 5 | 421 | 76 | |
BC009806.2 | - | 21 - 22 | 2189 | 76 | |
BC050420.1 | - | 21 - 22 | 2214 | 76 | |
J04031.1 | - | 21 - 22 | 2189 | 76 |