Assay Design



Gene Transcripts

Gene Symbol
MARK3
Entrez Gene ID
4140
Gene Name
microtubule affinity regulating kinase 3
Gene Aliases
CTAK1, KP78, PAR1A, Par-1a
Chromosome Location
Chr.14: 103385364 - 103503831 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128918.2 NP_001122390.2 10 - 11 1660 150
NM_001128919.2 NP_001122391.2 10 - 11 1660 150
NM_001128921.2 NP_001122393.2 9 - 10 1423 150
NM_002376.6 NP_002367.5 10 - 11 1660 150
XM_005267641.4 XP_005267698.2 10 - 11 1039 150
XM_005267643.4 XP_005267700.2 10 - 11 1660 150
XM_006720146.3 XP_006720209.1 10 - 11 1034 150
XM_011536769.2 XP_011535071.1 9 - 10 773 150
XM_017021294.1 XP_016876783.1 9 - 10 1409 150
XM_017021295.1 XP_016876784.1 9 - 10 1424 150
XM_017021296.1 XP_016876785.1 9 - 10 1441 150
XM_017021298.1 XP_016876787.1 9 - 10 1417 150
XM_017021300.1 XP_016876789.1 8 - 9 894 150
XM_017021301.1 XP_016876790.1 8 - 9 1049 150
XM_017021302.1 XP_016876791.1 8 - 9 894 150
XM_017021303.1 XP_016876792.1 8 - 9 894 150
XM_017021304.1 XP_016876793.1 8 - 9 894 150
XM_017021306.1 XP_016876795.1 9 - 10 1589 150
XM_017021307.1 XP_016876796.1 8 - 9 1540 150
XM_017021309.1 XP_016876798.1 8 - 9 872 150
XM_017021310.1 XP_016876799.1 8 - 9 872 150
XM_017021311.1 XP_016876800.1 7 - 8 823 150
GenBank mRNA AF387637.1 - 10 - 11 1012 150
AF465413.1 - 10 - 11 1504 150
AK307448.1 - 10 - 11 1039 150
BC024773.1 - 10 - 11 1483 150
BX161395.1 - 9 - 10 1353 150
U64205.1 - 10 - 11 1369 150

View Full Product Details