Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142471.1 | NP_001135943.1 | 3 - 4 | 404 | 128 |
NM_020395.3 | NP_065128.2 | 4 - 5 | 566 | 128 | |
XM_005263148.4 | XP_005263205.1 | 3 - 4 | 1059 | 128 | |
XM_011532143.2 | XP_011530445.1 | 4 - 5 | 1215 | 128 | |
XM_011532145.2 | XP_011530447.1 | 5 - 6 | 702 | 128 | |
GenBank mRNA | AF242524.1 | - | 3 - 4 | 398 | 128 |
AK314939.1 | - | 3 - 4 | 404 | 128 | |
BC014442.1 | - | 4 - 5 | 514 | 128 |