Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145128.2 | NP_001138600.2 | 25 - 26 | 3043 | 104 |
XM_006715376.3 | XP_006715439.2 | 26 - 27 | 3908 | 104 | |
XM_011535550.2 | XP_011533852.1 | 26 - 27 | 3908 | 104 | |
XM_011535552.2 | XP_011533854.1 | 25 - 26 | 3084 | 104 | |
XM_011535553.2 | XP_011533855.1 | 25 - 26 | 3691 | 104 | |
XM_011535554.2 | XP_011533856.1 | 24 - 25 | 3587 | 104 | |
XM_011535555.2 | XP_011533857.1 | 26 - 27 | 3908 | 104 | |
XM_011535556.2 | XP_011533858.1 | 26 - 27 | 3908 | 104 | |
XM_011535558.1 | XP_011533860.1 | 7 - 8 | 929 | 104 | |
XM_017010384.1 | XP_016865873.1 | 26 - 27 | 3908 | 104 | |
XM_017010385.1 | XP_016865874.1 | 25 - 26 | 3076 | 104 | |
XM_017010386.1 | XP_016865875.1 | 26 - 27 | 3908 | 104 | |
XM_017010387.1 | XP_016865876.1 | 20 - 21 | 3539 | 104 | |
XM_017010388.1 | XP_016865877.1 | 26 - 27 | 3908 | 104 | |
XM_017010389.1 | XP_016865878.1 | 11 - 12 | 1791 | 104 | |
GenBank mRNA | AK131244.1 | - | 1 - 2 | 451 | 104 |