Assay Design



Gene Transcripts

Gene Symbol
AK9
Entrez Gene ID
221264
Gene Name
adenylate kinase 9
Gene Aliases
AK 9, AKD1, AKD2, C6orf199, C6orf224, dJ70A9.1
Chromosome Location
Chr.6: 109492855 - 109691253 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145128.2 NP_001138600.2 32 - 33 4356 131
XM_006715376.3 XP_006715439.2 33 - 34 5221 131

View Full Product Details