Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001807.4 | NP_001798.2 | 1 - 2 | 88 | 90 |
GenBank mRNA | AK299965.1 | - | 1 - 2 | 66 | 90 |
BC042510.1 | - | 1 - 2 | 73 | 90 | |
M85201.1 | - | 1 - 2 | 80 | 90 | |
X54457.1 | - | 1 - 2 | 153 | 90 |