Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007259.2 | NP_001007260.1 | 6 - 7 | 892 | 104 |
NM_001007261.2 | NP_001007262.1 | 7 - 8 | 986 | 104 | |
NM_001286752.1 | NP_001273681.1 | 7 - 8 | 1375 | 104 | |
NM_017438.4 | NP_059134.1 | 6 - 7 | 1232 | 104 | |
NR_040087.2 | 5 - 6 | 1143 | 104 | ||
XM_011529636.1 | XP_011527938.1 | 6 - 7 | 1281 | 104 | |
XM_011529637.1 | XP_011527939.1 | 6 - 7 | 1346 | 104 | |
XM_011529638.2 | XP_011527940.1 | 6 - 7 | 1254 | 104 | |
XM_011529639.1 | XP_011527941.1 | 6 - 7 | 1032 | 104 | |
XM_011529640.2 | XP_011527942.1 | 6 - 7 | 1297 | 104 | |
XM_011529642.1 | XP_011527944.1 | 7 - 8 | 1375 | 104 | |
XM_011529643.1 | XP_011527945.1 | 7 - 8 | 1326 | 104 | |
XM_011529644.1 | XP_011527946.1 | 7 - 8 | 887 | 104 | |
XM_017028403.1 | XP_016883892.1 | 6 - 7 | 823 | 104 | |
XM_017028404.1 | XP_016883893.1 | 7 - 8 | 931 | 104 | |
XM_017028405.1 | XP_016883894.1 | 6 - 7 | 1232 | 104 | |
GenBank mRNA | AB004848.1 | - | 7 - 8 | 939 | 104 |
AB004849.1 | - | 1 - 2 | 442 | 104 | |
AB004852.1 | - | 5 - 6 | 763 | 104 | |
AB004853.1 | - | 7 - 8 | 935 | 104 | |
AF391112.1 | - | 5 - 6 | 710 | 104 | |
AK001660.1 | - | 6 - 7 | 799 | 104 | |
AK095215.1 | - | 5 - 6 | 2099 | 104 | |
AK300009.1 | - | 5 - 6 | 843 | 104 | |
BC002898.2 | - | 6 - 7 | 831 | 104 | |
BC036556.2 | - | 7 - 8 | 1049 | 104 |