Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145913.3 | NP_666018.3 | 6 - 7 | 1220 | 85 |
XM_017018910.1 | XP_016874399.1 | 6 - 7 | 1256 | 85 | |
GenBank mRNA | AF536216.1 | - | 6 - 7 | 1193 | 85 |
AK075263.1 | - | 6 - 7 | 878 | 85 | |
AK313788.1 | - | 6 - 7 | 1189 | 85 | |
AY081220.1 | - | 6 - 7 | 1194 | 85 | |
BC110492.2 | - | 6 - 7 | 855 | 85 |