Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152489.1 | NP_689702.1 | 3 - 4 | 492 | 153 |
XM_005270498.4 | XP_005270555.1 | 3 - 4 | 1286 | 153 | |
XM_006710379.3 | XP_006710442.1 | 3 - 4 | 427 | 153 | |
XM_011540764.2 | XP_011539066.1 | 4 - 5 | 1227 | 153 | |
GenBank mRNA | BC029895.1 | - | 3 - 4 | 492 | 153 |