Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_206808.3 | NP_996531.1 | 5 - 6 | 670 | 96 |
NR_104592.1 | 5 - 6 | 670 | 96 | ||
XM_005254030.2 | XP_005254087.1 | 5 - 6 | 649 | 96 | |
XM_005254031.2 | XP_005254088.1 | 4 - 5 | 547 | 96 | |
XM_006719915.3 | XP_006719978.1 | 5 - 6 | 649 | 96 | |
XM_011521051.2 | XP_011519353.1 | 5 - 6 | 649 | 96 | |
XM_011521052.2 | XP_011519354.1 | 5 - 6 | 649 | 96 | |
XM_017020402.1 | XP_016875891.1 | 4 - 5 | 547 | 96 | |
XM_017020403.1 | XP_016875892.1 | 4 - 5 | 1368 | 96 | |
XM_017020404.1 | XP_016875893.1 | 4 - 5 | 1517 | 96 | |
GenBank mRNA | AF428253.1 | - | 5 - 6 | 638 | 96 |
AK095506.1 | - | 5 - 6 | 647 | 96 | |
AK300510.1 | - | 4 - 5 | 563 | 96 | |
BC034360.1 | - | 5 - 6 | 652 | 96 |