Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018321.3 | NP_060791.3 | 2 - 3 | 291 | 154 |
GenBank mRNA | AF347667.1 | - | 2 - 3 | 267 | 154 |
AK001962.1 | - | 2 - 3 | 296 | 154 | |
AK289610.1 | - | 2 - 3 | 290 | 154 | |
AK303311.1 | - | 2 - 3 | 330 | 154 | |
AY364243.1 | - | 2 - 3 | 267 | 154 | |
AY364244.1 | - | 2 - 3 | 267 | 154 | |
BC001546.1 | - | 2 - 3 | 291 | 154 | |
BC006014.1 | - | 2 - 3 | 294 | 154 | |
BC036741.1 | - | 2 - 3 | 641 | 154 |