Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018321.3 | NP_060791.3 | 6 - 7 | 529 | 108 |
GenBank mRNA | AF347667.1 | - | 6 - 7 | 505 | 108 |
AK289610.1 | - | 6 - 7 | 528 | 108 | |
AK307492.1 | - | 2 - 3 | 112 | 108 | |
AY364244.1 | - | 6 - 7 | 505 | 108 | |
BC001546.1 | - | 6 - 7 | 529 | 108 | |
BC006014.1 | - | 6 - 7 | 532 | 108 | |
BC036741.1 | - | 6 - 7 | 879 | 108 |