Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190818.1 | NP_001177747.1 | 11 - 12 | 1983 | 68 |
NM_001190819.1 | NP_001177748.1 | 11 - 12 | 1971 | 68 | |
NM_004153.3 | NP_004144.2 | 11 - 12 | 1986 | 68 | |
XM_011541527.2 | XP_011539829.1 | 11 - 12 | 1935 | 68 | |
XM_017001389.1 | XP_016856878.1 | 12 - 13 | 2023 | 68 | |
GenBank mRNA | AK291909.1 | - | 11 - 12 | 1961 | 68 |
BC011539.1 | - | 11 - 12 | 1899 | 68 | |
U43416.1 | - | 11 - 12 | 1966 | 68 |