Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190818.1 | NP_001177747.1 | 3 - 4 | 443 | 66 |
NM_001190819.1 | NP_001177748.1 | 3 - 4 | 446 | 66 | |
NM_004153.3 | NP_004144.2 | 3 - 4 | 446 | 66 | |
XM_011541527.2 | XP_011539829.1 | 3 - 4 | 338 | 66 | |
XM_017001388.1 | XP_016856877.1 | 3 - 4 | 446 | 66 | |
XM_017001389.1 | XP_016856878.1 | 3 - 4 | 421 | 66 | |
GenBank mRNA | AK291909.1 | - | 3 - 4 | 421 | 66 |
AK303420.1 | - | 3 - 4 | 387 | 66 | |
BC011539.1 | - | 3 - 4 | 359 | 66 | |
BP280383.1 | - | 3 - 4 | 443 | 66 | |
DB156841.1 | - | 3 - 4 | 446 | 66 | |
U40152.1 | - | 3 - 4 | 434 | 66 | |
U43416.1 | - | 3 - 4 | 426 | 66 |