Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003568.2 | NP_003559.2 | 5 - 6 | 737 | 67 |
XM_011510058.2 | XP_011508360.1 | 5 - 6 | 456 | 67 | |
GenBank mRNA | AF230929.1 | - | 5 - 6 | 493 | 67 |
AJ009985.1 | - | 5 - 6 | 682 | 67 | |
AK314926.1 | - | 5 - 6 | 456 | 67 | |
BC005830.2 | - | 4 - 5 | 393 | 67 | |
CR536481.1 | - | 3 - 4 | 246 | 67 |