Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003568.2 | NP_003559.2 | 7 - 8 | 939 | 113 |
XM_011510058.2 | XP_011508360.1 | 7 - 8 | 658 | 113 | |
GenBank mRNA | AF230929.1 | - | 7 - 8 | 695 | 113 |
AJ009985.1 | - | 7 - 8 | 884 | 113 | |
AK314926.1 | - | 7 - 8 | 658 | 113 | |
BC005830.2 | - | 6 - 7 | 595 | 113 | |
CR536481.1 | - | 5 - 6 | 448 | 113 |