Assay Design



Gene Transcripts

Gene Symbol
OVGP1
Entrez Gene ID
5016
Gene Name
oviductal glycoprotein 1
Gene Aliases
CHIT5, EGP, MUC9, OGP
Chromosome Location
Chr.1: 111414315 - 111427777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002557.3 NP_002548.3 2 - 3 117 66
GenBank mRNA AK295207.1 - 1 - 2 457 66
AK314074.1 - 2 - 3 75 66
AY189737.1 - 2 - 3 118 66
BC126177.1 - 2 - 3 100 66
BC136406.1 - 2 - 3 100 66
U09550.1 - 2 - 3 73 66

View Full Product Details