Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002557.3 | NP_002548.3 | 6 - 7 | 665 | 102 |
GenBank mRNA | AB208783.1 | - | 4 - 5 | 1047 | 102 |
AK301726.1 | - | 2 - 3 | 188 | 102 | |
AK314074.1 | - | 6 - 7 | 623 | 102 | |
AY189737.1 | - | 6 - 7 | 666 | 102 | |
BC126177.1 | - | 6 - 7 | 648 | 102 | |
BC136406.1 | - | 6 - 7 | 648 | 102 | |
U09550.1 | - | 6 - 7 | 621 | 102 |