Assay Design



Gene Transcripts

Gene Symbol
OVGP1
Entrez Gene ID
5016
Gene Name
oviductal glycoprotein 1
Gene Aliases
CHIT5, EGP, MUC9, OGP
Chromosome Location
Chr.1: 111414315 - 111427777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002557.3 NP_002548.3 7 - 8 771 71
GenBank mRNA AB208783.1 - 5 - 6 1153 71
AK298757.1 - 5 - 6 514 71
AK301726.1 - 3 - 4 294 71
AK314074.1 - 7 - 8 729 71
AY189737.1 - 7 - 8 772 71
BC126177.1 - 7 - 8 754 71
BC136406.1 - 7 - 8 754 71
U09550.1 - 7 - 8 727 71

View Full Product Details