Assay Design



Gene Transcripts

Gene Symbol
PRTG
Entrez Gene ID
283659
Gene Name
protogenin
Gene Aliases
IGDCC5
Chromosome Location
Chr.15: 55611540 - 55743649 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173814.5 NP_776175.2 7 - 8 1319 131
XM_011521459.1 XP_011519761.1 7 - 8 1849 131
XM_011521460.1 XP_011519762.1 7 - 8 1849 131
XM_017022081.1 XP_016877570.1 7 - 8 1406 131
XM_017022082.1 XP_016877571.1 7 - 8 1849 131
GenBank mRNA AY630258.1 - 7 - 8 1179 131

View Full Product Details