Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000761.4 | NP_000752.2 | 3 - 4 | 1014 | 74 |
GenBank mRNA | AF182274.1 | - | 2 - 3 | 74 | |
BC067424.1 | - | 3 - 4 | 994 | 74 | |
BC067425.1 | - | 3 - 4 | 994 | 74 | |
BC067426.1 | - | 3 - 4 | 994 | 74 | |
BC067427.1 | - | 3 - 4 | 994 | 74 | |
BC067428.1 | - | 3 - 4 | 994 | 74 | |
BC067429.1 | - | 3 - 4 | 994 | 74 | |
M12078.1 | - | 1 - 2 | 68 | 74 | |
M55053.1 | - | 2 - 3 | 958 | 74 | |
Z00036.1 | - | 2 - 3 | 958 | 74 |