Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303135.1 | NP_001290064.1 | 2 - 3 | 74 | 72 |
NM_006229.3 | NP_006220.1 | 2 - 3 | 84 | 72 | |
XM_011539868.1 | XP_011538170.1 | 2 - 3 | 217 | 72 | |
XM_011539869.1 | XP_011538171.1 | 2 - 3 | 71 | 72 | |
GenBank mRNA | AK130236.1 | - | 2 - 3 | 72 | 72 |
AK307063.1 | - | 2 - 3 | 71 | 72 | |
BC005233.1 | - | 2 - 3 | 84 | 72 | |
BC025784.2 | - | 1 - 2 | 47 | 72 | |
BP323340.1 | - | 2 - 3 | 74 | 72 | |
CR749524.1 | - | 2 - 3 | 83 | 72 | |
M93283.1 | - | 2 - 3 | 66 | 72 |