Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099271.1 | NP_001092741.1 | 11 - 12 | 1787 | 79 |
NM_152408.2 | NP_689621.2 | 10 - 11 | 1586 | 79 | |
XM_005248436.1 | XP_005248493.1 | 11 - 12 | 1703 | 79 | |
XM_011543159.1 | XP_011541461.1 | 12 - 13 | 1793 | 79 | |
XM_011543160.1 | XP_011541462.1 | 10 - 11 | 1647 | 79 | |
XM_017009037.1 | XP_016864526.1 | 10 - 11 | 1682 | 79 | |
XM_017009038.1 | XP_016864527.1 | 10 - 11 | 1707 | 79 | |
XM_017009039.1 | XP_016864528.1 | 10 - 11 | 1626 | 79 | |
XM_017009040.1 | XP_016864529.1 | 9 - 10 | 1485 | 79 | |
GenBank mRNA | AK093098.1 | - | 10 - 11 | 1556 | 79 |
AK296071.1 | - | 11 - 12 | 1704 | 79 | |
BC101325.1 | - | 10 - 11 | 1552 | 79 | |
BC101326.2 | - | 10 - 11 | 1556 | 79 | |
BC101327.2 | - | 10 - 11 | 1556 | 79 | |
CA441610.1 | - | 1 - 2 | 79 |