Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099271.1 | NP_001092741.1 | 9 - 10 | 1320 | 102 |
NM_152408.2 | NP_689621.2 | 8 - 9 | 1119 | 102 | |
XM_011543158.2 | XP_011541460.1 | 9 - 10 | 1320 | 102 | |
XM_011543159.1 | XP_011541461.1 | 10 - 11 | 1326 | 102 | |
XM_011543160.1 | XP_011541462.1 | 8 - 9 | 1180 | 102 | |
XM_017009037.1 | XP_016864526.1 | 8 - 9 | 1215 | 102 | |
XM_017009038.1 | XP_016864527.1 | 8 - 9 | 1240 | 102 | |
XM_017009039.1 | XP_016864528.1 | 8 - 9 | 1159 | 102 | |
XM_017009040.1 | XP_016864529.1 | 7 - 8 | 1018 | 102 | |
GenBank mRNA | AK093098.1 | - | 8 - 9 | 1089 | 102 |
AK296071.1 | - | 9 - 10 | 1237 | 102 | |
BC101325.1 | - | 8 - 9 | 1085 | 102 | |
BC101326.2 | - | 8 - 9 | 1089 | 102 | |
BC101327.2 | - | 8 - 9 | 1089 | 102 | |
BC101328.2 | - | 8 - 9 | 1086 | 102 |