Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002334.3 | NP_002325.2 | 22 - 23 | 3382 | 69 |
XM_011520103.2 | XP_011518405.1 | 16 - 17 | 2452 | 69 | |
XM_011520104.2 | XP_011518406.1 | 7 - 8 | 919 | 69 | |
XM_017017734.1 | XP_016873223.1 | 22 - 23 | 3299 | 69 | |
GenBank mRNA | AB011540.1 | - | 14 - 15 | 2152 | 69 |
AB084910.1 | - | 22 - 23 | 3382 | 69 | |
AB231861.1 | - | 22 - 23 | 3285 | 69 | |
BC136667.1 | - | 22 - 23 | 3303 | 69 | |
BC136668.1 | - | 22 - 23 | 3303 | 69 |