Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002334.3 | NP_002325.2 | 33 - 34 | 5196 | 69 |
XM_011520103.2 | XP_011518405.1 | 27 - 28 | 4266 | 69 | |
XM_011520104.2 | XP_011518406.1 | 18 - 19 | 2733 | 69 | |
XM_017017734.1 | XP_016873223.1 | 33 - 34 | 5113 | 69 | |
GenBank mRNA | AB011540.1 | - | 25 - 26 | 3966 | 69 |
AB084910.1 | - | 33 - 34 | 5196 | 69 | |
AB231861.1 | - | 33 - 34 | 5090 | 69 | |
BC136667.1 | - | 33 - 34 | 5117 | 69 | |
BC136668.1 | - | 33 - 34 | 5117 | 69 | |
DB061460.1 | - | 3 - 4 | 381 | 69 |