Assay Design



Gene Transcripts

Gene Symbol
UBP1
Entrez Gene ID
7342
Gene Name
upstream binding protein 1 (LBP-1a)
Gene Aliases
LBP-1B, LBP-1a, LBP1A, LBP1B
Chromosome Location
Chr.3: 33388336 - 33440405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128160.1 NP_001121632.1 12 - 13 1817 79
NM_001128161.1 NP_001121633.1 14 - 15 1544 79
NM_014517.4 NP_055332.3 13 - 14 1925 79
GenBank mRNA AF198487.1 - 13 - 14 1569 79
AK023274.1 - 12 - 13 1446 79
AK023278.1 - 7 - 8 727 79
AK293100.1 - 14 - 15 1544 79
AK309430.1 - 1 - 2 465 79
BC047235.1 - 13 - 14 1924 79
CR749798.1 - 1 - 2 2282 79

View Full Product Details