Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128160.1 | NP_001121632.1 | 5 - 6 | 1086 | 69 |
NM_001128161.1 | NP_001121633.1 | 6 - 7 | 705 | 69 | |
NM_014517.4 | NP_055332.3 | 5 - 6 | 1086 | 69 | |
GenBank mRNA | AF198487.1 | - | 5 - 6 | 730 | 69 |
AK023274.1 | - | 5 - 6 | 715 | 69 | |
AK293100.1 | - | 6 - 7 | 705 | 69 | |
BC047235.1 | - | 5 - 6 | 1085 | 69 |