Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128161.1 | NP_001121633.1 | 9 - 10 | 1078 | 80 |
NM_014517.4 | NP_055332.3 | 8 - 9 | 1459 | 80 | |
GenBank mRNA | AF198487.1 | - | 8 - 9 | 1103 | 80 |
AK293100.1 | - | 9 - 10 | 1078 | 80 | |
BC047235.1 | - | 8 - 9 | 1458 | 80 |