Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003009.2 | NP_003000.1 | 2 - 3 | 254 | 101 |
GenBank mRNA | AF015283.1 | - | 2 - 3 | 103 | 101 |
BC000581.2 | - | 2 - 3 | 119 | 101 | |
BC032546.1 | - | 2 - 3 | 144 | 101 | |
BC039597.1 | - | 2 - 3 | 122 | 101 | |
BC047893.1 | - | 2 - 3 | 121 | 101 | |
BF973456.1 | - | 2 - 3 | 267 | 101 | |
U67171.1 | - | 2 - 3 | 114 | 101 |