Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002593.3 | NP_002584.2 | 1 - 2 | 298 | 99 |
GenBank mRNA | AB008549.1 | - | 1 - 2 | 156 | 99 |
AK298364.1 | - | 1 - 2 | 156 | 99 | |
AK313748.1 | - | 1 - 2 | 194 | 99 | |
BC000574.2 | - | 1 - 2 | 191 | 99 | |
BC033205.1 | - | 1 - 2 | 136 | 99 | |
BI837971.1 | - | 1 - 2 | 298 | 99 | |
L33799.1 | - | 1 - 2 | 160 | 99 |