Assay Design



Gene Transcripts

Gene Symbol
EPHB6
Entrez Gene ID
2051
Gene Name
EPH receptor B6
Gene Aliases
HEP
Chromosome Location
Chr.7: 142854640 - 142871093 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001280795.2 NP_001267724.2 1 - 2 79 125
GenBank mRNA BC110606.1 - 1 - 2 76 125

View Full Product Details