Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004445.5 | NP_004436.4 | 3 - 4 | 348 | 103 |
NR_104001.2 | 4 - 5 | 465 | 103 | ||
XM_006715881.2 | XP_006715944.1 | 6 - 7 | 1343 | 103 | |
XM_011515879.1 | XP_011514181.1 | 7 - 8 | 1382 | 103 | |
XM_011515880.1 | XP_011514182.1 | 6 - 7 | 1360 | 103 | |
XM_011515881.2 | XP_011514183.1 | 6 - 7 | 1344 | 103 | |
XM_011515882.2 | XP_011514184.1 | 6 - 7 | 1346 | 103 | |
GenBank mRNA | BC110607.1 | - | 3 - 4 | 345 | 103 |
BX410199.2 | - | 2 - 3 | 270 | 103 | |
D83492.1 | - | 6 - 7 | 922 | 103 | |
EU054308.1 | - | 2 - 3 | 124 | 103 |